Assay Details
| Gene Symbol: Fzr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019757.1 | NP_062731.1 | 4 - 5 | 403 | 57 | |
| GenBank mRNA | AF083809.1 | - | 4 - 5 | 403 | 57 | |
| AK051993.1 | - | 4 - 5 | 485 | 57 | ||
| AK081476.1 | - | 4 - 5 | 457 | 57 | ||
| AK083686.1 | - | 466 | 57 | |||
| AK154822.1 | - | 4 - 5 | 487 | 57 | ||
| AK155370.1 | - | 4 - 5 | 485 | 57 | ||
| AK163951.1 | - | 4 - 5 | 462 | 57 | ||
| BC006616.1 | - | 4 - 5 | 484 | 3967009 | 57 | |
More Information
Set Membership: |
|

Fzr1
Genomic Map