Assay Details
| Gene Symbol: Ick | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163780.1 | NP_001157252.1 | 9 - 10 | 1607 | 89 | |
| NM_019987.2 | NP_064371.2 | 9 - 10 | 1535 | 89 | ||
| GenBank mRNA | AF225918.1 | - | 9 - 10 | 1375 | 89 | |
| AK052178.1 | - | 2 - 3 | 504 | 89 | ||
| AK133537.1 | - | 9 - 10 | 1474 | 89 | ||
| AK154185.1 | - | 9 - 10 | 1608 | 89 | ||
| AK173068.1 | - | 4 - 5 | 764 | 89 | ||
| BC028863.1 | - | 8 - 9 | 1292 | 4224269 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ick
Genomic Map