Assay Details
| Gene Symbol: Fibp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253832.1 | NP_001240761.1 | 1 - 2 | 168 | 71 | |
| NM_021438.4 | NP_067413.1 | 1 - 2 | 168 | 71 | ||
| GenBank mRNA | AF270700.1 | - | 1 - 2 | 81 | 71 | |
| AK008093.1 | - | 1 - 2 | 159 | 71 | ||
| AK008132.1 | - | 1 - 2 | 154 | 71 | ||
| AK012629.1 | - | 1 - 2 | 151 | 71 | ||
| AK019399.1 | - | 1 - 2 | 139 | 71 | ||
| AK028029.1 | - | 1 - 2 | 159 | 71 | ||
| AK090037.1 | - | 1 - 2 | 194 | 71 | ||
| BC031716.1 | - | 1 - 2 | 138 | 3969355 | 71 | |
| BC049284.1 | - | 1 - 2 | 146 | 6395407 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fibp
Genomic Map