Assay Details
| Gene Symbol: Fibp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253832.1 | NP_001240761.1 | 8 - 9 | 1010 | 90 | |
| NM_021438.4 | NP_067413.1 | 8 - 9 | 989 | 90 | ||
| GenBank mRNA | AF270700.1 | - | 8 - 9 | 902 | 90 | |
| AK008132.1 | - | 8 - 9 | 993 | 90 | ||
| AK012629.1 | - | 8 - 9 | 972 | 90 | ||
| AK019399.1 | - | 8 - 9 | 981 | 90 | ||
| AK028029.1 | - | 8 - 9 | 980 | 90 | ||
| BC031716.1 | - | 8 - 9 | 980 | 3969355 | 90 | |
| BC049284.1 | - | 8 - 9 | 988 | 6395407 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518246 | BC031716.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518246 | BC049284.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518246 | AK028029.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518246 | AF270700.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518246 | AK008132.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518246 | BC031716.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518246 | BC049284.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518246 | AK028029.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518246 | AF270700.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518246 | AK008132.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815535 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 76248 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 47061 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162694 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518246 | BC031716.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518246 | BC049284.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518246 | AK028029.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518246 | AF270700.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518246 | AK008132.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518246 | BC031716.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518246 | BC049284.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518246 | AK028029.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518246 | AF270700.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518246 | AK008132.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928826 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928816 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928808 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31928814 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162694 | - | - |
| Whole Mouse Genome G4122A | A_51_P518246 | BC031716.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518246 | BC049284.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518246 | AK028029.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518246 | AF270700.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518246 | AK008132.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518246 | BC031716.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518246 | BC049284.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518246 | AK028029.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518246 | AF270700.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518246 | AK008132.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P427992 | BC031716.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P427992 | BC049284.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P427992 | AK028029.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P427992 | AK019399.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P427992 | AK012629.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125301 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120648 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162694 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14180164 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14180171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31928812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14247029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928800 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14180167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928803 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928824 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928827 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928808 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928802 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928826 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928830 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928834 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14180153 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928816 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928811 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14372844 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928829 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928835 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928840 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31928812 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10460598 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460602 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460596 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460599 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460597 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460595 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460600 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460592 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460594 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460593 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460601 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430171_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438924_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422516_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438923_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10460591 | - | - |
| Mouse Genome 430 2.0 Array | 1438923_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422516_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1438923_at | - | - |
| Mouse Genome 430 2.0 Array | 1438924_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1430171_at | - | - |
| Mouse Genome 430 2.0 Array | 1438924_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1430171_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422516_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 58249_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 58249_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900000843.mm.1 | NM_001253832.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000843.mm.1 | NM_001253832.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000843.mm.1 | NM_021438.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000843.mm.1 | NM_021438.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000843.mm.1 | TC1900000069.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900000841.mm.1 | NM_001253832.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000841.mm.1 | NM_001253832.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000841.mm.1 | NM_021438.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000841.mm.1 | NM_021438.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000841.mm.1 | TC1900000069.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422516_a_at | - | - |
| Mouse Expression Set 430 | 1438924_x_at | - | - |
| Mouse Expression Set 430 | 1438923_PM_at | - | - |
| Mouse Expression Set 430 | 1438924_PM_x_at | - | - |
| Mouse Expression Set 430 | 1422516_PM_a_at | - | - |
| Mouse Expression Set 430 | 1438923_at | - | - |
| Murine Genome U74v2 Set | 99176_at | - | - |
| Murine Genome U74v2 Set | 99176_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422516_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438924_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438923_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422516_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438923_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1438924_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430171_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438924_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438923_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422516_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430171_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438923_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422516_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1438924_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102030338 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2510450 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102030338 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002510450 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1248068 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1260261 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1248068 | - | - |
| mouse-6 v1.1 (Illumina) | scl0058249.2_204-S | - | - |
| mouse-6 v1.1 (Illumina) | scl52842.4.1_43-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102030338 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2510450 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0058249.2_204-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2510450 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102030338 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2674132 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1260261 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2674132 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0058249.2_204-S | - | - |

Fibp
Genomic Map