Assay Details
| Gene Symbol: Impg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022016.3 | NP_071299.3 | 11 - 12 | 1365 | 91 | |
| GenBank mRNA | AF229929.1 | - | 11 - 12 | 1276 | 91 | |
| AF266478.1 | - | 11 - 12 | 1411 | 91 | ||
| BC022970.1 | - | 11 - 12 | 1353 | 5366008 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Impg1
Genomic Map