Assay Details
| Gene Symbol: Impg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022016.3 | NP_071299.3 | 13 - 14 | 1992 | 88 | |
| GenBank mRNA | AF229929.1 | - | 13 - 14 | 1903 | 88 | |
| AF266478.1 | - | 13 - 14 | 2038 | 88 | ||
| BC022970.1 | - | 13 - 14 | 1980 | 5366008 | 88 | |
More Information
Set Membership: |
|

Impg1
Genomic Map