| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900022712.mm.1 |
NM_022016.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900022712.mm.1 |
NM_022016.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900022712.mm.1 |
TC0900002745.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900022711.mm.1 |
NM_022016.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900022711.mm.1 |
NM_022016.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900022711.mm.1 |
TC0900002745.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900022713.mm.1 |
NM_022016.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900022713.mm.1 |
NM_022016.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900022713.mm.1 |
TC0900002745.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900022710.mm.1 |
NM_022016.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900022710.mm.1 |
NM_022016.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900022710.mm.1 |
TC0900002745.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900022714.mm.1 |
NM_022016.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900022714.mm.1 |
NM_022016.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900022714.mm.1 |
TC0900002745.mm.1 |
MTA TC |