Assay Details
| Gene Symbol: Bhmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022884.2 | NP_075022.2 | 6 - 7 | 807 | 71 | |
| GenBank mRNA | AF257474.1 | - | 6 - 7 | 793 | 71 | |
| AK090308.1 | - | 6 - 7 | 785 | 71 | ||
| AK145901.1 | - | 6 - 7 | 808 | 71 | ||
| BC013515.1 | - | 6 - 7 | 804 | 4235455 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
methionine biosynthetic process
methylation
S-methylmethionine metabolic process
S-methylmethionine cycle
S-adenosylmethionine metabolic process
L-methionine salvage
methyltransferase activity
zinc ion binding
S-adenosylmethionine-homocysteine S-methyltransferase activity
transferase activity
metal ion binding
betaine-homocysteine S-methyltransferase activity
S-methylmethionine-homocysteine S-methyltransferase activity
methionine biosynthetic process
methylation
S-methylmethionine metabolic process
S-methylmethionine cycle
S-adenosylmethionine metabolic process
L-methionine salvage
methyltransferase activity
zinc ion binding
S-adenosylmethionine-homocysteine S-methyltransferase activity
transferase activity
metal ion binding
betaine-homocysteine S-methyltransferase activity
S-methylmethionine-homocysteine S-methyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321734 | BC013515.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321734 | AF257474.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321734 | AK145901.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321734 | NM_022884.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321734 | BC013515.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321734 | AF257474.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321734 | AK145901.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P321734 | NM_022884.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 716024 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 99293 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 301497 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 166831 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 705961 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 54234 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 744902 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 259445 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 227157 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 727513 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 651524 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 157649 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 55760 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119966 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102779 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321734 | BC013515.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321734 | AF257474.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321734 | AK145901.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321734 | NM_022884.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321734 | BC013515.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321734 | AF257474.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321734 | AK145901.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P321734 | NM_022884.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321734 | BC013515.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321734 | AF257474.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321734 | AK145901.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321734 | NM_022884.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321734 | BC013515.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321734 | AF257474.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321734 | AK145901.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P321734 | NM_022884.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29230401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29230408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29230427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29230422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29230412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29230425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29230426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29230414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29230400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29230413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29230416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29230405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29230411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29230421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29230399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29230423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29230419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29230407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29230397 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102779 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07955962 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119966 | - | - |
| Whole Mouse Genome G4122A | A_51_P321734 | BC013515.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321734 | AF257474.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321734 | AK145901.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321734 | NM_022884.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321734 | BC013515.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321734 | AF257474.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321734 | AK145901.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P321734 | NM_022884.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110093 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119966 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102779 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10987814 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10987816 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11121237 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10765855 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10670261 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10879918 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10987818 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11121241 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10670263 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11121234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29230391 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10673427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230407 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10565119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230390 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230414 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10835690 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230403 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230399 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10835701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10673430 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230416 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10448086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230397 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230423 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10465632 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10835696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230420 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10938372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10826183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230391 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230422 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29230421 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10411148 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10457116 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411151 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411150 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411152 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411155 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411154 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411149 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411153 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418913_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418914_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10411147 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10511886 | - | - |
| Mouse Genome 430 2.0 Array | 1418914_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1418914_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1418913_at | - | - |
| Mouse Genome 430 2.0 Array | 1418913_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 64918_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 64918_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300016269.mm.1 | NM_022884.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300016269.mm.1 | NM_022884.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300016269.mm.1 | TC1300002437.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300016270.mm.1 | NM_022884.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300016270.mm.1 | NM_022884.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300016270.mm.1 | TC1300002437.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300016271.mm.1 | NM_022884.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300016271.mm.1 | NM_022884.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300016271.mm.1 | TC1300002437.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418914_s_at | - | - |
| Mouse Expression Set 430 | 1418914_PM_s_at | - | - |
| Mouse Expression Set 430 | 1418913_at | - | - |
| Mouse Expression Set 430 | 1418913_PM_at | - | - |
| Murine Genome U74v2 Set | 97418_PM_at | - | - |
| Murine Genome U74v2 Set | 97418_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418913_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418914_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418914_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418913_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418913_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418914_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418914_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418913_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 910974 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2030184 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002030184 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2651019 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2651019 | - | - |
| mouse-6 v1.1 (Illumina) | scl43677.9_377-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2030184 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl43677.9_377-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2030184 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2651019 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2651019 | - | - |

Bhmt2
Genomic Map