Assay Details
| Gene Symbol: Arid5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023598.2 | NP_076087.2 | 3 - 4 | 516 | 70 | |
| GenBank mRNA | AF169968.1 | - | 741 | 70 | ||
| AF280065.1 | - | 3 - 4 | 530 | 70 | ||
| AK017396.1 | - | 3 - 4 | 517 | 70 | ||
| AK029818.1 | - | 3 - 4 | 512 | 70 | ||
| AK031596.1 | - | 3 - 4 | 512 | 70 | ||
| AK136201.1 | - | 2 - 3 | 242 | 70 | ||
| BY717211.1 | - | 3 - 4 | 517 | 70 | ||
| BY720203.1 | - | 2 - 3 | 242 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arid5b
Genomic Map