Assay Details
| Gene Symbol: Gpn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024216.1 | NP_077178.1 | 7 - 8 | 904 | 89 | |
| GenBank mRNA | AK016996.1 | - | 7 - 8 | 835 | 89 | |
| AK042405.1 | - | 1619 | 89 | |||
| AK080720.1 | - | 7 - 8 | 851 | 89 | ||
| AK087968.1 | - | 7 - 8 | 855 | 89 | ||
| AK146783.1 | - | 7 - 8 | 842 | 89 | ||
| BC003341.1 | - | 7 - 8 | 904 | 3257412 | 89 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24050010 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224445 | AK146783.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224445 | BC003341.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224445 | AK016996.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224445 | AK042405.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224445 | AK080720.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224445 | AK146783.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224445 | BC003341.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224445 | AK016996.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224445 | AK042405.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224445 | AK080720.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 237614 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 438275 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 381806 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 870377 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 684460 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 197449 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 101336 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104768 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114186 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224445 | AK146783.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224445 | BC003341.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224445 | AK016996.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224445 | AK042405.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224445 | AK080720.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224445 | AK146783.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224445 | BC003341.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224445 | AK016996.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224445 | AK042405.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224445 | AK080720.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050019 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05724439 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104768 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05724451 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114186 | - | - |
| Whole Mouse Genome G4122A | A_51_P224445 | AK146783.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224445 | BC003341.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224445 | AK016996.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224445 | AK042405.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224445 | AK080720.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224445 | AK146783.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224445 | BC003341.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224445 | AK016996.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224445 | AK042405.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224445 | AK080720.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131643 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132205 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104768 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114186 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04914226 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04725138 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24050010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050019 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24050038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04694688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24050032 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04540881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04699357 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24050023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24050029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04694683 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24050026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04491159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04699358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24050008 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04699356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04725138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04637384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04699355 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04314141 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24050014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04694689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04314142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24050025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24050027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24050021 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04645907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04914226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24050024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04491158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04914235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24050013 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10525396 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525395 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525390 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525392 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525391 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525393 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525389 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525394 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424682_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10525387 | - | - |
| Mouse Genome 430 2.0 Array | 1424682_at | - | - |
| Mouse Genome 430 2.0 Array | 1424682_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68080_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68080_at | - | - |
| Mouse Expression Set 430 | 1424682_PM_at | - | - |
| Mouse Expression Set 430 | 1424682_at | - | - |
| Murine Genome U74v2 Set | 103884_at | - | - |
| Murine Genome U74v2 Set | 103884_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424682_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424682_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424682_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424682_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102100168 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102100168 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4610484 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004610484 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2745005 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234607 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2745005 | - | - |
| mouse-6 v1.1 (Illumina) | scl0068080.2_0-S | - | - |
| mouse-6 v1.1 (Illumina) | scl27258.6_57-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102100168 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4610484 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0068080.2_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102100168 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4610484 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2745005 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2745005 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1234607 | - | - |

Gpn3
Genomic Map