Assay Details
| Gene Symbol: Anapc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025514.2 | NP_079790.1 | 2 - 3 | 328 | 59 | |
| GenBank mRNA | AK009172.1 | - | 2 - 3 | 328 | 59 | |
| AK012280.1 | - | 2 - 3 | 196 | 59 | ||
| AK088376.1 | - | 2 - 3 | 276 | 59 | ||
| BC025117.1 | - | 321 | 3984181 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27171932 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27171936 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27171933 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27171935 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333071 | NM_025514.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333071 | AK009172.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333071 | NM_025514.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P333071 | AK009172.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 353372 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649347 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610179 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 807850 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 677495 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506797 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 436646 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 389185 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336769 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 528829 | AK077727.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 528829 | AK162212.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 528829 | NM_001024952.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 119898 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 136369 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165123 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137099 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333071 | NM_025514.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333071 | AK009172.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333071 | NM_025514.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P333071 | AK009172.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27171948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27171932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27171944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27171940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27171955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27171951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27171937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27171957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27171942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27171933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27171936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27171956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27171938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27171946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27171941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27171959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27171958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27171935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27171953 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02135229 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165123 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137099 | - | - |
| Whole Mouse Genome G4122A | A_51_P333071 | NM_025514.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333071 | AK009172.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333071 | NM_025514.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P333071 | AK009172.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P550684 | AK037565.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138301 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137099 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165123 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08242352 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08242364 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08242356 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08242367 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08242354 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08242357 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27171936 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27171932 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27171935 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27171933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27171869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08637473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171840 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08314599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08569205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08541267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08242364 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08314612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08588519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08316972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08894973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08637470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171857 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08493843 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08493848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08460142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08588522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27171976 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10369299 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369296 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369300 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369297 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10369298 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449338_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449339_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419995_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419994_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10369295 | - | - |
| Mouse Genome 430 2.0 Array | 1419994_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1419994_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1419995_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449339_at | - | - |
| Mouse Genome 430 2.0 Array | 1419995_at | - | - |
| Mouse Genome 430 2.0 Array | 1449338_at | - | - |
| Mouse Genome 430 2.0 Array | 1449339_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449338_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52717_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52717_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000015540.mm.1 | NM_025514.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000015540.mm.1 | TC1000002218.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449338_PM_at | - | - |
| Mouse Expression Set 430 | 1449338_at | - | - |
| Mouse Expression Set 430 | 1419995_PM_at | - | - |
| Mouse Expression Set 430 | 1449339_at | - | - |
| Mouse Expression Set 430 | 1419994_PM_s_at | - | - |
| Mouse Expression Set 430 | 1419995_at | - | - |
| Mouse Expression Set 430 | 1449339_PM_at | - | - |
| Mouse Expression Set 430 | 1419994_s_at | - | - |
| Murine Genome U74v2 Set | 104087_PM_at | - | - |
| Murine Genome U74v2 Set | 104087_at | - | - |
| Murine Genome U74v2 Set | 169287_PM_i_at | - | - |
| Murine Genome U74v2 Set | 169287_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449338_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419994_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419994_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419995_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449339_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419995_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449339_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449338_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449339_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419994_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449339_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419994_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419995_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449338_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419995_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449338_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 587951 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3440138 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003440138 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1219088 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1219088 | - | - |
| mouse-6 v1.1 (Illumina) | scl37933.6_401-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3440138 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl37933.6_401-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3440138 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1219088 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1219088 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl37933.6_401-S | - | - |

Anapc16
Genomic Map