Assay Details
| Gene Symbol: Ankzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267620.1 | NP_001254549.1 | 2 - 3 | 625 | 68 | |
| NM_026187.5 | NP_080463.3 | 2 - 3 | 625 | 68 | ||
| NR_051998.1 | 2 - 3 | 625 | 68 | |||
| GenBank mRNA | AK004948.2 | - | 2 - 3 | 593 | 68 | |
| AK147915.1 | - | 1 - 2 | 2136 | 68 | ||
| AK156255.1 | - | 2 - 3 | 589 | 68 | ||
| AK157405.1 | - | 2 - 3 | 589 | 68 | ||
| BC039910.1 | - | 2 - 3 | 588 | 68 | ||
| BC051449.1 | - | 2 - 3 | 605 | 68 | ||
| BC071240.1 | - | 2 - 3 | 524 | 68 | ||
| BC150879.1 | - | 2 - 3 | 575 | 9087790 | 68 | |
| DV062487.1 | - | 2 - 3 | 649 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK156255.1 transcripts.
Set Membership: |
|

Ankzf1
Genomic Map