Assay Details
| Gene Symbol: Ankzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026187.5 | NP_080463.3 | 5 - 6 | 1036 | 68 | |
| NR_051998.1 | 5 - 6 | 1067 | 68 | |||
| GenBank mRNA | AK004948.2 | - | 3 - 4 | 1466 | 68 | |
| AK156255.1 | - | 5 - 6 | 1000 | 68 | ||
| AK157405.1 | - | 5 - 6 | 1000 | 68 | ||
| BC039910.1 | - | 4 - 5 | 1342 | 68 | ||
| BC071240.1 | - | 5 - 6 | 966 | 68 | ||
| BC150879.1 | - | 5 - 6 | 986 | 9087790 | 68 | |
| BU054541.1 | - | 4 - 5 | 437 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK156255.1 transcripts.
Set Membership: |
|

Ankzf1
Genomic Map