Assay Details
| Gene Symbol: Mpnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026530.5 | NP_080806.4 | 4 - 5 | 672 | 88 | |
| GenBank mRNA | AK012128.1 | - | 3 - 4 | 627 | 88 | |
| AK146973.1 | - | 4 - 5 | 657 | 88 | ||
| AK160356.1 | - | 4 - 5 | 641 | 88 | ||
| AK172381.1 | - | 3 - 4 | 415 | 88 | ||
| AK213160.1 | - | 2 - 3 | 161 | 88 | ||
| BC017625.1 | - | 3 - 4 | 621 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31253272 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31253269 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31253271 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31253270 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31253273 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017112 | BC017625.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017112 | AK146973.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017112 | AK160356.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017112 | AK172381.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017112 | AK212460.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505322 | BC017625.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505322 | AK146973.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505322 | AK172381.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505322 | NM_026530.5 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505322 | AK012128.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505322 | BC017625.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505322 | AK146973.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505322 | AK172381.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505322 | NM_026530.5 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505322 | AK012128.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 927602 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 437416 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 158502 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 421147 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 190339 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 74625 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017112 | BC017625.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017112 | AK146973.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017112 | AK160356.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017112 | AK172381.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017112 | AK212460.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505322 | BC017625.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505322 | AK146973.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505322 | AK172381.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505322 | NM_026530.5 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505322 | AK012128.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505322 | BC017625.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505322 | AK146973.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505322 | AK172381.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505322 | NM_026530.5 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505322 | AK012128.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253273 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31253275 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08810155 | - | - |
| Whole Mouse Genome G4122A | A_51_P505322 | BC017625.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505322 | AK146973.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505322 | AK172381.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505322 | NM_026530.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505322 | AK012128.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505322 | BC017625.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505322 | AK146973.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505322 | AK172381.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505322 | NM_026530.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505322 | AK012128.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108586 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08810155 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13300380 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31253273 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31253269 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31253270 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31253271 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31253272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31253270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13238573 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13370158 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13439078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13089024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253275 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13092953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253308 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13238568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253310 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253299 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253281 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253265 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13092949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253273 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253279 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13089023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13089025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253282 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31253266 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10446015 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446023 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446014 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446019 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446026 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446025 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446024 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446017 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446018 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446022 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446020 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446021 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446016 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451357_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10446013 | - | - |
| Mouse Genome 430 2.0 Array | 1451357_at | - | - |
| Mouse Genome 430 2.0 Array | 1451357_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68047_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68047_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700009494.mm.1 | NM_026530.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009494.mm.1 | NM_026530.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009494.mm.1 | TC1700000996.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700009493.mm.1 | NM_026530.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009493.mm.1 | NM_026530.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009493.mm.1 | TC1700000996.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451357_PM_at | - | - |
| Mouse Expression Set 430 | 1451357_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451357_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451357_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451357_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451357_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 465758 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2190017 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2810167 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002190017 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002810167 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2597052 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2730803 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2730803 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2597052 | - | - |
| mouse-6 v1.1 (Illumina) | scl0001652.1_18-S | - | - |
| mouse-6 v1.1 (Illumina) | scl50604.11.1_29-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2190017 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2810167 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001652.1_18-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50604.11.1_29-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2810167 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2190017 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2730803 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2730803 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl50604.11.1_29-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001652.1_18-S | - | - |

Mpnd
Genomic Map