Assay Details
| Gene Symbol: Cpsf4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026682.2 | NP_080958.2 | 4 - 5 | 433 | 72 | |
| NM_029794.2 | NP_084070.1 | 4 - 5 | 489 | 72 | ||
| GenBank mRNA | AK002449.1 | - | 3 - 4 | 72 | ||
| AK005072.1 | - | 4 - 5 | 489 | 72 | ||
| BC016190.1 | - | 4 - 5 | 427 | 4483305 | 72 | |
| DV659633.1 | - | 4 - 5 | 423 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cpsf4l
Genomic Map