Assay Details
| Gene Symbol: Esam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027102.3 | NP_081378.1 | 5 - 6 | 873 | 80 | |
| GenBank mRNA | AF361882.1 | - | 5 - 6 | 858 | 80 | |
| AK009223.1 | - | 5 - 6 | 848 | 80 | ||
| AK156399.1 | - | 5 - 6 | 871 | 80 | ||
| BC049249.1 | - | 5 - 6 | 853 | 5039892 | 80 | |
| BG077536.2 | - | 3 - 4 | 307 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Esam
Genomic Map