Assay Details
| Gene Symbol: Ceacam13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027210.3 | NP_081486.1 | 3 - 4 | 866 | 79 | |
| GenBank mRNA | AK005430.1 | - | 3 - 4 | 857 | 79 | |
| AK146045.1 | - | 3 - 4 | 867 | 79 | ||
| BC099430.1 | - | 3 - 4 | 853 | 30787167 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ceacam13
Genomic Map