Assay Details
| Gene Symbol: Slc52a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029643.3 | NP_083919.1 | 1 - 2 | 248 | 74 | |
| GenBank mRNA | AK008081.1 | - | 1 - 2 | 227 | 74 | |
| AK029579.1 | - | 1 - 2 | 249 | 74 | ||
| AK032015.1 | - | 1 - 2 | 228 | 74 | ||
| AK080449.1 | - | 1 - 2 | 322 | 74 | ||
| AK142597.1 | - | 1 - 2 | 330 | 74 | ||
| AK152335.1 | - | 1 - 2 | 254 | 74 | ||
| AK161447.1 | - | 1 - 2 | 360 | 74 | ||
| BC016264.1 | - | 1 - 2 | 308 | 4921900 | 74 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK008081.1,AK029579.1,AK032015.1,AK080449.1,AK142597.1,AK152335.1,AK161447.1,BC016264.1,NM_029643.3 transcripts.
Set Membership: |
|

Slc52a2
Genomic Map