Assay Details
| Gene Symbol: Ptgr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252625.1 | NP_001239554.1 | 7 - 8 | 1140 | 85 | |
| NM_001252626.1 | NP_001239555.1 | 8 - 9 | 1212 | 85 | ||
| NM_029880.3 | NP_084156.2 | 7 - 8 | 1083 | 85 | ||
| GenBank mRNA | AK007522.1 | - | 8 - 9 | 1096 | 85 | |
| AK020666.1 | - | 7 - 8 | 1017 | 85 | ||
| AK021033.1 | - | 7 - 8 | 975 | 85 | ||
| AK036168.1 | - | 7 - 8 | 1005 | 85 | ||
| AK145232.1 | - | 7 - 8 | 1031 | 85 | ||
| AK159187.1 | - | 7 - 8 | 979 | 85 | ||
| AK159932.1 | - | 7 - 8 | 980 | 85 | ||
| AK168895.1 | - | 7 - 8 | 974 | 85 | ||
| BC021466.1 | - | 7 - 8 | 1029 | 5068419 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptgr2
Genomic Map