Assay Details
| Gene Symbol: Creb3l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB047647.1 | - | 6 - 7 | 655 | 94 | |
| AB052779.1 | - | 8 - 9 | 912 | 94 | ||
| AB182651.1 | - | 8 - 9 | 1030 | 94 | ||
| AB182652.1 | - | 6 - 7 | 655 | 94 | ||
| AF287260.1 | - | 6 - 7 | 793 | 94 | ||
| AK005918.1 | - | 6 - 7 | 812 | 94 | ||
| AK019928.1 | - | 8 - 9 | 1030 | 94 | ||
| AK088919.1 | - | 8 - 9 | 929 | 94 | ||
| AY049978.1 | - | 8 - 9 | 951 | 94 | ||
| BC022605.1 | - | 8 - 9 | 886 | 4164994 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Creb3l4
Genomic Map