Assay Details
| Gene Symbol: Cnnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031396.2 | NP_113573.2 | 4 - 5 | 2037 | 64 | |
| GenBank mRNA | AF202994.1 | - | 4 - 5 | 1263 | 64 | |
| DQ885890.5 | - | 4 - 5 | 933 | 64 | ||
| EF041508.1 | - | 4 - 5 | 933 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cnnm1
Genomic Map