Assay Details
| Gene Symbol: Wwc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133791.4 | NP_598552.2 | 8 - 9 | 1314 | 113 | |
| GenBank mRNA | AB045323.1 | - | 8 - 9 | 1337 | 113 | |
| AK032989.1 | - | 4 - 5 | 380 | 113 | ||
| AK079375.1 | - | 8 - 9 | 1149 | 113 | ||
| AK086956.1 | - | 8 - 9 | 1144 | 113 | ||
| AK147648.1 | - | 1308 | 113 | |||
| AK147944.1 | - | 8 - 9 | 1283 | 113 | ||
| AK209117.1 | - | 2 - 3 | 200 | 113 | ||
| AK211421.1 | - | 3 - 4 | 262 | 113 | ||
| BC067050.1 | - | 8 - 9 | 1314 | 30537299 | 113 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB045323.1 transcripts.
Set Membership: |
|

Wwc2
Genomic Map