Assay Details
| Gene Symbol: Exd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133798.3 | NP_598559.2 | 7 - 8 | 1605 | 69 | |
| GenBank mRNA | AK146901.1 | - | 7 - 8 | 1486 | 69 | |
| AK147922.1 | - | 7 - 8 | 1466 | 69 | ||
| AK158201.1 | - | 7 - 8 | 1496 | 69 | ||
| AK162206.1 | - | 6 - 7 | 1373 | 69 | ||
| AK164329.1 | - | 7 - 8 | 1481 | 69 | ||
| AK167692.1 | - | 7 - 8 | 1606 | 69 | ||
| BC018508.1 | - | 6 - 7 | 1148 | 3156447 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exd2
Genomic Map