Assay Details
| Gene Symbol: Dhx32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286030.1 | NP_001272959.1 | 9 - 10 | 2178 | 110 | |
| NM_001286031.1 | NP_001272960.1 | 9 - 10 | 2047 | 110 | ||
| NM_001286032.1 | NP_001272961.1 | 8 - 9 | 1617 | 110 | ||
| NM_133941.2 | NP_598702.1 | 9 - 10 | 2131 | 110 | ||
| GenBank mRNA | AK028910.1 | - | 9 - 10 | 1988 | 110 | |
| AK033624.1 | - | 9 - 10 | 2133 | 110 | ||
| AK079184.1 | - | 8 - 9 | 1517 | 110 | ||
| AK135760.1 | - | 9 - 10 | 2181 | 110 | ||
| AK167889.1 | - | 9 - 10 | 2076 | 110 | ||
| AK169005.1 | - | 9 - 10 | 2122 | 110 | ||
| AK172504.1 | - | 9 - 10 | 2112 | 110 | ||
| AK195868.1 | - | 1 - 2 | 88 | 110 | ||
| AY065980.1 | - | 9 - 10 | 2133 | 110 | ||
| BC006911.1 | - | 8 - 9 | 1688 | 110 | ||
| BC022920.1 | - | 8 - 9 | 1887 | 4460064 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhx32
Genomic Map