Assay Details
| Gene Symbol: Dhx32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286030.1 | NP_001272959.1 | 10 - 11 | 2362 | 88 | |
| NM_001286031.1 | NP_001272960.1 | 10 - 11 | 2231 | 88 | ||
| NM_001286032.1 | NP_001272961.1 | 9 - 10 | 1801 | 88 | ||
| NM_133941.2 | NP_598702.1 | 10 - 11 | 2315 | 88 | ||
| GenBank mRNA | AK028910.1 | - | 10 - 11 | 2172 | 88 | |
| AK033624.1 | - | 10 - 11 | 2317 | 88 | ||
| AK079184.1 | - | 9 - 10 | 1701 | 88 | ||
| AK167889.1 | - | 10 - 11 | 2260 | 88 | ||
| AK169005.1 | - | 10 - 11 | 2306 | 88 | ||
| AK172504.1 | - | 10 - 11 | 2296 | 88 | ||
| AY065980.1 | - | 10 - 11 | 2317 | 88 | ||
| BC006911.1 | - | 9 - 10 | 1872 | 88 | ||
| BC022920.1 | - | 9 - 10 | 2071 | 4460064 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhx32
Genomic Map