Assay Details
| Gene Symbol: Cog4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133973.2 | NP_598734.1 | 9 - 10 | 1212 | 61 | |
| GenBank mRNA | AK156466.1 | - | 9 - 10 | 1203 | 61 | |
| AK172465.1 | - | 9 - 10 | 1203 | 61 | ||
| AK191447.1 | - | 1 - 2 | 50 | 61 | ||
| AK199943.1 | - | 1 - 2 | 50 | 61 | ||
| AY223866.1 | - | 9 - 10 | 1239 | 61 | ||
| BC023120.1 | - | 9 - 10 | 1189 | 4161858 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cog4
Genomic Map