Assay Details
| Gene Symbol: Cog4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133973.2 | NP_598734.1 | 10 - 11 | 1334 | 105 | |
| GenBank mRNA | AK172465.1 | - | 10 - 11 | 1325 | 105 | |
| AK191447.1 | - | 2 - 3 | 172 | 105 | ||
| AY223866.1 | - | 10 - 11 | 1361 | 105 | ||
| BC023120.1 | - | 10 - 11 | 1311 | 4161858 | 105 | |
More Information
Set Membership: |
|

Cog4
Genomic Map