Assay Details
| Gene Symbol: Pla2g16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139269.2 | NP_644675.2 | 2 - 3 | 176 | 79 | |
| GenBank mRNA | AK039703.1 | - | 2 - 3 | 389 | 79 | |
| AK042007.1 | - | 2 - 3 | 129 | 79 | ||
| AK052657.1 | - | 2 - 3 | 155 | 79 | ||
| AK163505.1 | - | 2 - 3 | 177 | 79 | ||
| AX775891.1 | - | 2 - 3 | 158 | 79 | ||
| BC024581.1 | - | 2 - 3 | 122 | 5065247 | 79 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK039703.1,AK042007.1,AK052657.1,AK163505.1,AX775891.1,BC024581.1,NM_139269.2 transcripts.
Set Membership: |
|

Pla2g16
Genomic Map