Assay Details
| Gene Symbol: Kif18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139303.1 | NP_647464.1 | 5 - 6 | 897 | 76 | |
| GenBank mRNA | AK036722.1 | - | 5 - 6 | 784 | 76 | |
| AK041469.1 | - | 5 - 6 | 796 | 76 | ||
| AK044795.1 | - | 5 - 6 | 792 | 76 | ||
| AK049197.1 | - | 5 - 6 | 847 | 76 | ||
| AK082913.1 | - | 5 - 6 | 810 | 76 | ||
| AK135735.1 | - | 5 - 6 | 818 | 76 | ||
| AK164649.1 | - | 5 - 6 | 792 | 76 | ||
| BC004740.1 | - | 4 - 5 | 752 | 76 | ||
| BC016095.1 | - | 5 - 6 | 897 | 4506052 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kif18a
Genomic Map