Assay Details
| Gene Symbol: Kif18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139303.1 | NP_647464.1 | 8 - 9 | 1350 | 117 | |
| GenBank mRNA | AK036722.1 | - | 8 - 9 | 1237 | 117 | |
| AK041469.1 | - | 8 - 9 | 1249 | 117 | ||
| AK044795.1 | - | 8 - 9 | 1245 | 117 | ||
| AK049197.1 | - | 8 - 9 | 1300 | 117 | ||
| AK164649.1 | - | 8 - 9 | 1245 | 117 | ||
| BC004740.1 | - | 7 - 8 | 1205 | 117 | ||
| BC016095.1 | - | 8 - 9 | 1350 | 4506052 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kif18a
Genomic Map