Assay Details
| Gene Symbol: Spice1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144550.4 | NP_653133.3 | 10 - 11 | 1265 | 72 | |
| GenBank mRNA | AK048789.1 | - | 10 - 11 | 1209 | 72 | |
| AY099108.1 | - | 10 - 11 | 1545 | 72 | ||
| BC024956.1 | - | 2 - 3 | 619 | 72 | ||
| BC052381.1 | - | 10 - 11 | 1219 | 5703283 | 72 | |
| BC056960.1 | - | 10 - 11 | 1219 | 5703283 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spice1
Genomic Map