Assay Details
| Gene Symbol: Spice1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144550.4 | NP_653133.3 | 16 - 17 | 2550 | 72 | |
| GenBank mRNA | AK048789.1 | - | 16 - 17 | 2494 | 72 | |
| AY099108.1 | - | 16 - 17 | 2830 | 72 | ||
| BC024956.1 | - | 8 - 9 | 1904 | 72 | ||
| BC052381.1 | - | 16 - 17 | 2504 | 5703283 | 72 | |
| BC056960.1 | - | 16 - 17 | 2504 | 5703283 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spice1
Genomic Map