Assay Details
| Gene Symbol: Lincpint | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110469.1 | 2 - 3 | 474 | 70 | ||
| GenBank mRNA | BC145649.1 | - | 2 - 3 | 424 | 70 | |
| KC860257.1 | - | 2 - 3 | 419 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984253 | BC145649.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984253 | AB041803.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2033557 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984259 | AB041803.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984259 | AB041803.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984253 | BC145649.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984253 | AB041803.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2033557 | - | - |
| Whole Mouse Genome G4122A | A_52_P26265 | AB041803.1 | mouse_transcriptome |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05578486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05211864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05578480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336418 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05578481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05433146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05046207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05695453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336081 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05846933 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05398546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05578478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05398553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05398533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05695455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05695457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05046205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05575736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05578477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24336182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05578488 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10543786 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10543787 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10543790 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10543788 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10543789 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460417_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10543785 | - | - |
| Mouse Genome 430 2.0 Array | 1458698_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1458698_at | - | - |
| Mouse Genome 430 2.0 Array | 1460417_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1460417_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 232685_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 232685_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600015898.mm.1 | NR_110469.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600015898.mm.1 | NR_110470.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600015898.mm.1 | TC0600002013.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1460417_at | - | - |
| Mouse Expression Set 430 | 1460417_PM_at | - | - |
| Murine Genome U74v2 Set | 134833_PM_at | - | - |
| Murine Genome U74v2 Set | 134833_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460417_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460417_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460417_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460417_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1214368 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1214368 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1214368 | - | - |

Lincpint
Genomic Map