Assay Details
| Gene Symbol: Lmcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144799.2 | NP_659048.1 | 2 - 3 | 303 | 65 | |
| GenBank mRNA | AK075847.1 | - | 2 - 3 | 304 | 65 | |
| AK159946.1 | - | 2 - 3 | 254 | 65 | ||
| BC019124.1 | - | 2 - 3 | 254 | 5007525 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lmcd1
Genomic Map