| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100028805.mm.1 |
NM_001160215.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028805.mm.1 |
NM_001160215.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028805.mm.1 |
NM_145141.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028805.mm.1 |
NM_145141.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028805.mm.1 |
NR_027666.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028805.mm.1 |
NR_027666.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028805.mm.1 |
TC0100003520.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100028804.mm.1 |
NM_001160215.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028804.mm.1 |
NM_001160215.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028804.mm.1 |
NM_145141.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028804.mm.1 |
NM_145141.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028804.mm.1 |
NR_027666.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028804.mm.1 |
NR_027666.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028804.mm.1 |
TC0100003520.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100028808.mm.1 |
NR_027666.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028808.mm.1 |
NR_027666.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028808.mm.1 |
TC0100003520.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100028803.mm.1 |
NM_001160215.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028803.mm.1 |
NM_001160215.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028803.mm.1 |
NM_145141.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028803.mm.1 |
NM_145141.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028803.mm.1 |
NR_027666.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028803.mm.1 |
NR_027666.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028803.mm.1 |
TC0100003520.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100028809.mm.1 |
NM_001160215.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028809.mm.1 |
NM_001160215.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028809.mm.1 |
NM_145141.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028809.mm.1 |
NM_145141.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028809.mm.1 |
NR_027666.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028809.mm.1 |
NR_027666.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100028809.mm.1 |
TC0100003520.mm.1 |
MTA TC |