Assay Details
| Gene Symbol: Nsun2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145354.5 | NP_663329.3 | 16 - 17 | 1895 | 102 | |
| GenBank mRNA | AK030124.1 | - | 15 - 16 | 1780 | 102 | |
| AK075999.1 | - | 6 - 7 | 674 | 102 | ||
| AK150631.1 | - | 16 - 17 | 1849 | 102 | ||
| AK151917.1 | - | 16 - 17 | 1849 | 102 | ||
| AK154789.1 | - | 16 - 17 | 1887 | 102 | ||
| AK171399.1 | - | 16 - 17 | 1852 | 102 | ||
| BC013625.1 | - | 12 - 13 | 1367 | 4011674 | 102 | |
| BC023735.1 | - | 10 - 11 | 1164 | 102 | ||
| DQ490066.1 | - | 16 - 17 | 1858 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nsun2
Genomic Map