Assay Details
| Gene Symbol: Sectm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145373.2 | NP_663348.2 | 3 - 4 | 523 | 78 | |
| GenBank mRNA | BC010462.1 | - | 3 - 4 | 490 | 3585040 | 78 |
More Information
Set Membership: |
|

Sectm1a
Genomic Map