Assay Details
| Gene Symbol: Lactb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145381.2 | NP_663356.1 | 3 - 4 | 470 | 68 | |
| GenBank mRNA | AK088932.1 | - | 3 - 4 | 483 | 68 | |
| AK090091.1 | - | 3 - 4 | 499 | 68 | ||
| AK143912.1 | - | 3 - 4 | 449 | 68 | ||
| BC004045.1 | - | 3 - 4 | 470 | 3496398 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lactb2
Genomic Map