Assay Details
| Gene Symbol: Tapbpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145391.2 | NP_663366.2 | 1 - 2 | 325 | 79 | |
| GenBank mRNA | BC017613.1 | - | 1 - 2 | 325 | 4912570 | 79 |
| BC094656.1 | - | 1 - 2 | 296 | 79 | ||
| BX517366.1 | - | 1 - 2 | 178 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tapbpl
Genomic Map