Assay Details
| Gene Symbol: Tapbpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145391.2 | NP_663366.2 | 5 - 6 | 1491 | 70 | |
| GenBank mRNA | BC017613.1 | - | 5 - 6 | 1491 | 4912570 | 70 |
| BC094656.1 | - | 5 - 6 | 1461 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505795 | BC094656.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505795 | BC017613.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505795 | NM_145391.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505795 | BC094656.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505795 | BC017613.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P505795 | NM_145391.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 785433 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 74047 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901005 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473689 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491872 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 5573 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126716 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505795 | BC094656.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505795 | BC017613.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505795 | NM_145391.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505795 | BC094656.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505795 | BC017613.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P505795 | NM_145391.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24819492 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01438535 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126716 | - | - |
| Whole Mouse Genome G4122A | A_51_P505795 | BC094656.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505795 | BC017613.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505795 | NM_145391.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505795 | BC094656.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505795 | BC017613.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P505795 | NM_145391.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121997 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126716 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05269402 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05269405 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05269412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24819529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05277509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05396982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05857876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05269408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05269402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05124388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05482564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05634272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819469 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05124392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05494283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05269405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05482565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05634273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24819484 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10547984 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547978 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547981 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547979 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547980 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547983 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10547982 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451544_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424961_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10547976 | - | - |
| Mouse Genome 430 2.0 Array | 1424961_at | - | - |
| Mouse Genome 430 2.0 Array | 1424961_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451544_at | - | - |
| Mouse Genome 430 2.0 Array | 1451544_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 213233_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 213233_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600024530.mm.1 | NM_145391.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600024530.mm.1 | NM_145391.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600024530.mm.1 | TC0600003139.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451544_at | - | - |
| Mouse Expression Set 430 | 1451544_PM_at | - | - |
| Mouse Expression Set 430 | 1424961_at | - | - |
| Mouse Expression Set 430 | 1424961_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451544_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451544_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424961_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424961_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451544_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451544_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424961_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424961_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 847591 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1770086 | - | - |
| MouseWG-6 v2 Expression BeadChip | 540519 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106900204 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1770086 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 540519 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001770086 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000540519 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2754866 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2606498 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2606498 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2754866 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236651 | - | - |
| mouse-6 v1.1 (Illumina) | scl0001043.1_38-S | - | - |
| mouse-6 v1.1 (Illumina) | scl28407.7.1_0-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_21703781-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 540519 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106900204 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1770086 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl28407.7.1_0-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001043.1_38-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106900204 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1770086 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 540519 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2606502 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2974611 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2606498 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2606502 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2974611 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2606498 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001043.1_38-S | - | - |

Tapbpl
Genomic Map