Assay Details
| Gene Symbol: Rnpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159624.1 | NP_001153096.1 | 8 - 9 | 1611 | 131 | |
| NM_145417.3 | NP_663392.2 | 9 - 10 | 1728 | 131 | ||
| GenBank mRNA | AK031138.1 | - | 8 - 9 | 1551 | 131 | |
| AK145466.1 | - | 9 - 10 | 1732 | 131 | ||
| AK159195.1 | - | 9 - 10 | 1671 | 131 | ||
| AK159445.1 | - | 9 - 10 | 1660 | 131 | ||
| AK159625.1 | - | 9 - 10 | 1671 | 131 | ||
| BC010520.1 | - | 9 - 10 | 1689 | 131 | ||
| BC019200.1 | - | 9 - 10 | 1699 | 5041005 | 131 | |
| Gene Symbol: Dnase1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK157889.1 | - | 131 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Rnpep
Genomic Map