Assay Details
| Gene Symbol: Cdc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285988.1 | NP_001272917.1 | 7 - 8 | 1066 | 128 | |
| NM_001285989.1 | NP_001272918.1 | 7 - 8 | 1066 | 128 | ||
| NM_001285990.1 | NP_001272919.1 | 6 - 7 | 968 | 128 | ||
| NM_145436.2 | NP_663411.2 | 7 - 8 | 1066 | 128 | ||
| GenBank mRNA | AK039377.1 | - | 7 - 8 | 863 | 128 | |
| AK134425.1 | - | 6 - 7 | 778 | 128 | ||
| AK148278.1 | - | 7 - 8 | 921 | 128 | ||
| AK161471.1 | - | 7 - 8 | 935 | 128 | ||
| BC023187.1 | - | 7 - 8 | 924 | 128 | ||
| BC157955.1 | - | 7 - 8 | 1065 | 9088128 | 128 | |
| BC172100.1 | - | 7 - 8 | 1065 | 9054386 | 128 | |
| BC172149.1 | - | 6 - 7 | 967 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdc27
Genomic Map