Assay Details
| Gene Symbol: Mfsd7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145447.2 | NP_663422.1 | 3 - 4 | 1336 | 61 | |
| GenBank mRNA | AK145696.1 | - | 3 - 4 | 1293 | 61 | |
| AY260574.1 | - | 3 - 4 | 1336 | 61 | ||
| AY260575.1 | - | 3 - 4 | 1336 | 61 | ||
| AY260576.1 | - | 3 - 4 | 1336 | 61 | ||
| BC011209.1 | - | 3 - 4 | 1336 | 4190027 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd7c
Genomic Map