Assay Details
| Gene Symbol: Oxnad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145460.2 | NP_663435.2 | 2 - 3 | 448 | 60 | |
| GenBank mRNA | AK028649.1 | - | 2 - 3 | 358 | 60 | |
| AK031978.1 | - | 1 - 2 | 77 | 60 | ||
| AK032160.1 | - | 2 - 3 | 365 | 60 | ||
| AK075924.1 | - | 1 - 2 | 75 | 60 | ||
| AK139884.1 | - | 2 - 3 | 448 | 60 | ||
| AK215115.1 | - | 1 - 2 | 106 | 60 | ||
| AK215185.1 | - | 1 - 2 | 106 | 60 | ||
| BC019806.1 | - | 2 - 3 | 226 | 4018387 | 60 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK031978.1 transcripts.
Set Membership: |
|

Oxnad1
Genomic Map