Assay Details
| Gene Symbol: Oxnad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145460.2 | NP_663435.2 | 4 - 5 | 621 | 79 | |
| GenBank mRNA | AK028649.1 | - | 4 - 5 | 531 | 79 | |
| AK031978.1 | - | 3 - 4 | 250 | 79 | ||
| AK032160.1 | - | 4 - 5 | 538 | 79 | ||
| AK075924.1 | - | 3 - 4 | 248 | 79 | ||
| AK139884.1 | - | 4 - 5 | 621 | 79 | ||
| AK215115.1 | - | 3 - 4 | 279 | 79 | ||
| AK215185.1 | - | 3 - 4 | 279 | 79 | ||
| BC019806.1 | - | 4 - 5 | 399 | 4018387 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Oxnad1
Genomic Map