Assay Details
| Gene Symbol: Nmrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145497.2 | NP_663472.1 | 8 - 9 | 679 | 71 | |
| GenBank mRNA | AK050579.1 | - | 8 - 9 | 710 | 71 | |
| AK169962.1 | - | 8 - 9 | 653 | 71 | ||
| BC016495.1 | - | 8 - 9 | 679 | 4221437 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31992145 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P663526 | AK050579.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P663526 | AK169962.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P663526 | BC016495.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P663526 | NM_145497.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P663526 | AK050579.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P663526 | AK169962.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P663526 | BC016495.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P663526 | NM_145497.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 606884 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350833 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 703349 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 507722 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 237445 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 443325 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719081 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 642134 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 929123 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 663308 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 66595 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 972660 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 137525 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 112180 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106163 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121195 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P663526 | AK050579.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P663526 | AK169962.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P663526 | BC016495.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P663526 | NM_145497.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P663526 | AK050579.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P663526 | AK169962.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P663526 | BC016495.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P663526 | NM_145497.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31992152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31992156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31992166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31992168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31992154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31992157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31992174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31992165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31992161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31992146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31992145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31992173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31992153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31992148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31992159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31992170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31992150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31992163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31992172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31992167 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121195 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106163 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09121605 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09121637 | - | - |
| Whole Mouse Genome G4122A | A_52_P663526 | AK050579.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P663526 | AK169962.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P663526 | BC016495.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P663526 | NM_145497.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P663526 | AK050579.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P663526 | AK169962.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P663526 | BC016495.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P663526 | NM_145497.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126779 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106163 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121195 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14368871 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14278279 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14368878 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31992145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31992213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992174 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14374381 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992145 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992161 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14168274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992153 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14473954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14153139 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992148 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14368872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992176 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14202746 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14168281 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992315 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992156 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992171 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992168 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14470726 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992146 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992175 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31992179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14168277 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10461910 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461912 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461913 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461914 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461919 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461916 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461918 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461911 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461917 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461920 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461915 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448048_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1447503_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425646_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10461909 | - | - |
| Mouse Genome 430 2.0 Array | 1425646_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1447503_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448048_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1447503_at | - | - |
| Mouse Genome 430 2.0 Array | 1448048_at | - | - |
| Mouse Genome 430 2.0 Array | 1425646_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 225994_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 225994_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900003222.mm.1 | NM_145497.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900003222.mm.1 | NM_145497.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900003222.mm.1 | TC1900000320.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900003223.mm.1 | NM_145497.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900003223.mm.1 | NM_145497.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900003223.mm.1 | TC1900000320.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900003220.mm.1 | NM_145497.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900003220.mm.1 | NM_145497.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900003220.mm.1 | TC1900000320.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425646_at | - | - |
| Mouse Expression Set 430 | 1448048_PM_at | - | - |
| Mouse Expression Set 430 | 1448048_at | - | - |
| Mouse Expression Set 430 | 1425646_PM_at | - | - |
| Mouse Expression Set 430 | 1447503_at | - | - |
| Mouse Expression Set 430 | 1447503_PM_at | - | - |
| Murine Genome U74v2 Set | 95920_PM_at | - | - |
| Murine Genome U74v2 Set | 95920_at | - | - |
| Murine Genome U74v2 Set | 166971_at | - | - |
| Murine Genome U74v2 Set | 166971_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425646_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448048_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448048_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425646_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425646_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448048_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425646_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448048_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447503_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447503_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 403690 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3780082 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003780082 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1242106 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1242106 | - | - |
| mouse-6 v1.1 (Illumina) | scl53305.8.1_17-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3780082 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53305.8.1_17-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3780082 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1242106 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1242106 | - | - |

Nmrk1
Genomic Map