Assay Details
| Gene Symbol: Pls3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166453.1 | NP_001159925.1 | 8 - 9 | 1023 | 76 | |
| NM_001166454.2 | NP_001159926.1 | 8 - 9 | 1018 | 76 | ||
| NM_145629.2 | NP_663604.1 | 8 - 9 | 1137 | 76 | ||
| GenBank mRNA | AB182243.1 | - | 7 - 8 | 892 | 76 | |
| AK146087.1 | - | 8 - 9 | 948 | 76 | ||
| AK146455.1 | - | 8 - 9 | 940 | 76 | ||
| AK160501.1 | - | 8 - 9 | 966 | 76 | ||
| AK165540.1 | - | 8 - 9 | 1025 | 76 | ||
| BC005459.1 | - | 8 - 9 | 1098 | 3495462 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pls3
Genomic Map