Assay Details
| Gene Symbol: Pls3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166453.1 | NP_001159925.1 | 9 - 10 | 1116 | 77 | |
| NM_001166454.2 | NP_001159926.1 | 9 - 10 | 1111 | 77 | ||
| NM_145629.2 | NP_663604.1 | 9 - 10 | 1230 | 77 | ||
| GenBank mRNA | AB182243.1 | - | 8 - 9 | 985 | 77 | |
| AK146087.1 | - | 9 - 10 | 1041 | 77 | ||
| AK146455.1 | - | 9 - 10 | 1033 | 77 | ||
| AK160501.1 | - | 9 - 10 | 1059 | 77 | ||
| BC005459.1 | - | 9 - 10 | 1191 | 3495462 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pls3
Genomic Map