| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200023017.mm.1 |
NM_001177706.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023017.mm.1 |
NM_001177706.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023017.mm.1 |
NM_001177707.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023017.mm.1 |
NM_001177707.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023017.mm.1 |
NM_001177708.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023017.mm.1 |
NM_001177708.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023017.mm.1 |
NM_001177709.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023017.mm.1 |
NM_001177709.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023017.mm.1 |
NM_001177710.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023017.mm.1 |
NM_001177710.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023017.mm.1 |
NM_145760.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023017.mm.1 |
NM_145760.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023017.mm.1 |
TC0200002811.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200023026.mm.1 |
NM_001177706.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023026.mm.1 |
NM_001177706.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023026.mm.1 |
NM_001177707.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023026.mm.1 |
NM_001177707.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023026.mm.1 |
NM_001177708.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023026.mm.1 |
NM_001177708.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023026.mm.1 |
NM_001177709.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023026.mm.1 |
NM_001177709.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023026.mm.1 |
NM_145760.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023026.mm.1 |
NM_145760.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023026.mm.1 |
TC0200002811.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200023025.mm.1 |
NM_001177706.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023025.mm.1 |
NM_001177706.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023025.mm.1 |
NM_001177707.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023025.mm.1 |
NM_001177707.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023025.mm.1 |
NM_001177708.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023025.mm.1 |
NM_001177708.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023025.mm.1 |
NM_001177709.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023025.mm.1 |
NM_001177709.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023025.mm.1 |
NM_001177710.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023025.mm.1 |
NM_001177710.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023025.mm.1 |
NM_145760.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023025.mm.1 |
NM_145760.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023025.mm.1 |
TC0200002811.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200023019.mm.1 |
NM_001177706.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023019.mm.1 |
NM_001177706.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023019.mm.1 |
NM_001177707.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023019.mm.1 |
NM_001177707.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023019.mm.1 |
TC0200002811.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200023018.mm.1 |
NM_001177706.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023018.mm.1 |
NM_001177706.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023018.mm.1 |
TC0200002811.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200023020.mm.1 |
NM_001177706.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023020.mm.1 |
NM_001177706.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023020.mm.1 |
NM_001177707.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023020.mm.1 |
NM_001177707.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023020.mm.1 |
NM_145760.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023020.mm.1 |
NM_145760.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200023020.mm.1 |
TC0200002811.mm.1 |
MTA TC |