Assay Details
| Gene Symbol: Fam129b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146119.2 | NP_666231.1 | 8 - 9 | 1190 | 74 | |
| GenBank mRNA | AK033735.1 | - | 8 - 9 | 1158 | 74 | |
| AK046583.1 | - | 8 - 9 | 1087 | 74 | ||
| AK051396.1 | - | 8 - 9 | 1129 | 74 | ||
| AK146953.1 | - | 8 - 9 | 1175 | 74 | ||
| AK149818.1 | - | 8 - 9 | 1101 | 74 | ||
| AK149888.1 | - | 8 - 9 | 1100 | 74 | ||
| AK150642.1 | - | 8 - 9 | 1128 | 74 | ||
| AK152207.1 | - | 8 - 9 | 1149 | 74 | ||
| AK153489.1 | - | 8 - 9 | 1134 | 74 | ||
| AK170512.1 | - | 8 - 9 | 1130 | 74 | ||
| BC027843.1 | - | 8 - 9 | 1110 | 5343013 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam129b
Genomic Map